Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBAS Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP177339 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177339 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177339 Supplier Novus Biologicals Supplier No. NBP177339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GBAS Polyclonal specifically detects GBAS in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GBAS
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. O75323
Gene Alias glioblastoma amplified sequence, Glioblastoma-amplified sequence, NIPSNAP2NipSnap2,4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2, protein NipSnap homolog 2
Gene Symbols NIPSNAP2
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the center of human NIPSNAP2. The immunogen is located within the first 50 amino acids of NIPSNAP2. Amino Acid Squence: RTWTSSSNRSREDS
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2631
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.