Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBL Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB104251 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB104251 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB104251 Supplier Novus Biologicals Supplier No. NBP298484AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GBL Polyclonal specifically detects GBL in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GBL
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias G protein beta subunit-like, Gable, GbetaL, GBLPop3, Lst8, LST8MGC111011, Mammalian lethal with SEC13 protein 8, mLST8, MTOR associated protein, LST8 homolog (S. cerevisiae), POP3, Protein GbetaL, target of rapamycin complex subunit LST8, TORC subunit LST8, WAT1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GBL fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 64223
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.