Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $404.50
Specifications
Antigen | GBP4 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145313
|
Invitrogen™
PA5145313 |
200 μL |
Each for $404.50
|
|
|||||
FEPA5145314
|
Invitrogen™
PA5145314 |
20 μL |
Each for $191.80
|
|
|||||
Description
Guanylate-binding proteins, such as GBP4, are induced by interferon and hydrolyze GTP to both GDP and GMP.[supplied by OMIM, Dec 2008].Specifications
GBP4 | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
Q61107 | |
17472, 310917 | |
Recombinant Mouse GBP4 protein, Met1-Cys304 (Accession #Q61107). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
AW228052; Gbp3; GBP4; GBP-4; GTP-binding protein 4; guanine nucleotide-binding protein 4; guanylate binding protein 4; guanylate nucleotide binding protein 3; guanylate nucleotide binding protein 4; guanylate-binding protein 4; guanylate-binding protein 4 homolog; macrophage activation 2; Mag-2; mKIAA4245; Mpa2; Mpa-2 | |
GBP4 | |
IgG | |
Protein A | |
GBP4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title