Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBP5 Antibody (OTI5C9), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N272343J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272343J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272343J646 Supplier Novus Biologicals Supplier No. NBP272343JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GBP5 Monoclonal specifically detects GBP5 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GBP5
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5C9
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GBP-5, GBP-TA antigen, GTP-binding protein 5, Guanine nucleotide-binding protein 5, guanylate binding protein 5, guanylate-binding protein 5
Gene Symbols GBP5
Host Species Mouse
Immunogen Full length recombinant protein of human GBP5 (NP_443174) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 115362.0
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.