Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBP6 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB163998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163998 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163998 Supplier Novus Biologicals Supplier No. NBP281719G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GBP6 Polyclonal antibody specifically detects GBP6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen GBP6
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias DKFZp686G0786, FLJ39135, GBP-6, GTP-binding protein 6, Guanine nucleotide-binding protein 6, guanylate binding protein family, member 6, guanylate-binding protein 6
Host Species Rabbit
Immunogen GBP6 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human GBP6. The immunogen is located within the last 50 amino acids of GBP6. Amino Acid Squence: VKGVSSLFKKHKLPF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 163351
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.