Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBX2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP243617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243617 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243617 Supplier Novus Biologicals Supplier No. NBP243617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects GBX2 in Human samples, and it is validated for Western Blotting

GBX2 Polyclonal specifically detects GBX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GBX2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS pH7, 1% BSA, 20% Glycerol with 0.025% Proclin 300
Gene Alias Gastrulation and brain-specific homeobox protein 2, gastrulation brain homeo box 2, gastrulation brain homeobox 2, homeobox protein GBX-2
Gene Symbols GBX2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GBX2. The exact sequence is proprietary.
Molecular Weight of Antigen 37 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2637
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.