Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GC 1
GREENER_CHOICE

Catalog No. p-4817102
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity thyroid receptor α (TRα) and TRβ agonist; thyromimetic

Thyromimetic; high affinity agonist at thyroid hormone receptor (TR) β and TRα receptors (KD values are 67 and 440 pM respectively). Displays 5- and 100-fold greater potency than the endogenous agonist T3 in vitro at TRα1 and TRβ1 receptors respectively. Promotes reverse cholesterol transport in chow- and fat-fed mice. Induces proliferation of hepatocytes and pancreatic acinar cells in rats independent of prior tissue injury.

Chemical Identifiers

CAS 211110-63-3
Molecular Formula C20H24O4

Specifications

Quantity 10 mg
Formula Weight 328.4
Purity >98%
Molecular Weight (g/mol) 328.408
Chemical Name or Material GC 1
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.