Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCAP1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen GCAP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15486820
SDP
View Documents
Novus Biologicals
NBP15486820UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP154868
SDP
View Documents
Novus Biologicals
NBP154868
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

GCAP1 Polyclonal specifically detects GCAP1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

GCAP1
Polyclonal
Rabbit
Vision
C6orf131, chromosome 6 open reading frame 131, CORD14, GCAP 1, GCAP1photoreceptor 1, GCAPGUCA, Guanylate cyclase activator 1A, guanylate cyclase activator 1A (retina), guanylin 1, retina, guanylyl cyclase-activating protein 1, GUCA1
GUCA1A
IgG
23 kDa
Western Blot
Unconjugated
RUO
P43080
2978
Synthetic peptides corresponding to GUCA1A(guanylate cyclase activator 1A (retina)) The peptide sequence was selected from the N terminal of GUCA1A. Peptide sequence LYEFRQFFGLKNLSPSASQYVEQMFETFDFNKDGYIDFMEYVAALSLVLK.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.