Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCAP2 Antibody (A1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB1205422AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1205422AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1205422AH Supplier Novus Biologicals Supplier No. NB1205422AF647

Mouse Monoclonal Antibody

GCAP2 Monoclonal specifically detects GCAP2 in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GCAP2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A1
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp686E1183, GCAP 2, GCAP2guanylate cyclase-activating protein, photoreceptor 2, Guanylate cyclase activator 1B, guanylate cyclase activator 1B (retina), guanylyl cyclase-activating protein 2, GUCA2, RP48
Gene Symbols GUCA1B
Host Species Mouse
Immunogen Bacterial expressed full length GCAP-2.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2979
Test Specificity This is specific for GCAP 2 and does not react with other isotypes.
Target Species Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.