Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB418870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB418870 25 μL
NBP183609 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB418870 Supplier Novus Biologicals Supplier No. NBP18360925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

GCC1 Polyclonal specifically detects GCC1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen GCC1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:20 - 1:50, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q96CN9
Gene Alias FLJ22035, GCC1P, GCC88Golgi coiled-coil protein 1, golgi coiled-coil 1, GRIP and coiled-coil domain containing 1, GRIP and coiled-coil domain-containing protein 1, MGC20706, peripheral membrane Golgi protein
Gene Symbols GCC1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:GDPADTSSSDSLTQALQLAAANEPTFFLYAEQLARKEVEITSLRKQKHRLEVEVHQLQDRLLEEGERHREEVAALQSHIEKNIRDQSREGANLEYLKNIIYRFLTLPDSLGRQQTLTAILTILHFSPEEKQVIMRLPT
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79571
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.