Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCC1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB125451 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB125451 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB125451 Supplier Novus Biologicals Supplier No. NBP310275100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GCC1 Polyclonal specifically detects GCC1 in Human samples. It is validated for Western Blot.

Specifications

Antigen GCC1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias FLJ22035, GCC1P, GCC88Golgi coiled-coil protein 1, golgi coiled-coil 1, GRIP and coiled-coil domain containing 1, GRIP and coiled-coil domain-containing protein 1, MGC20706, peripheral membrane Golgi protein
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C terminal region of human GCC1 (NP_078799.2). Peptide sequence EVHQLQDRLLEEGERHREEVAALQSHIEKNIRDQSREGANLEYLKNIIYR
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79571
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.