Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCH1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | GCH1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1797720
![]() |
Novus Biologicals
NBP17977120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179771
![]() |
Novus Biologicals
NBP179771 |
100 μL |
Each for $499.50
|
|
|||||
Description
GCH1 Polyclonal specifically detects GCH1 in Human samples. It is validated for Western Blot.Specifications
GCH1 | |
Polyclonal | |
Rabbit | |
NP_000152 | |
2643 | |
Synthetic peptide directed towards the C terminal of human GCH1The immunogen for this antibody is GCH1. Peptide sequence LRPAGVGVVVEATHMCMVMRGVQKMNSKTVTSTMLGVFREDPKTREEFLT. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
dystonia 14, DYT14, DYT5a, DYT5EC 3.5.4.16, GCH, GTP cyclohydrolase 1, GTP cyclohydrolase I, GTPCH1, GTP-CH-1, GTP-CH-I, guanosine 5'-triphosphate cyclohydrolase I, HPABH4B | |
GCH1 | |
IgG | |
28 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title