Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCLC Antibody (OTI1A3), PE, Novus Biologicals™
SDP

Catalog No. NB289337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289337 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289337 Supplier Novus Biologicals Supplier No. NBP270830PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GCLC Monoclonal antibody specifically detects GCLC in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen GCLC
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1A3
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias EC 6.3.2.2, gamma-ECS, Gamma-glutamylcysteine synthetase, GCS, GCS heavy chain, GLCL, GLCLCGCL, glutamate--cysteine ligase catalytic subunit, glutamate-cysteine ligase, catalytic subunit
Host Species Mouse
Immunogen Full length human recombinant protein of human GCLC(NP_001489) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2729
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.