Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCM2 Rabbit anti-Human, Janelia Fluor 646, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB056241 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056241 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056241 Supplier Novus Biologicals Supplier No. NBP298880JF646

Rabbit Polyclonal Antibody

GCM2 Polyclonal specifically detects GCM2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GCM2
Applications Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias chorion-specific transcription factor GCMb, GCM motif protein 2, GCMBglial cells missing homolog b, glial cells missing (Drosophila) homolog b, Glial cells missing homolog 2, glial cells missing homolog 2 (Drosophila), glide/gcm protein homolog, hGCMbglial cells missing 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human GCM2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9247
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.