Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCN1L1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10097851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10097851 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10097851 Supplier Novus Biologicals Supplier No. NB10097851
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GCN1L1 Polyclonal specifically detects GCN1L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen GCN1L1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation 2-10 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q92616
Gene Alias GCN1, GCN1 (general control of amino-acid synthesis 1, yeast)-like 1, GCN1 general control of amino-acid synthesis 1-like 1 (yeast), GCN1L, GCN1-like protein 1, HsGCN1, KIAA0219hsGCN1, peroxisome proliferator activated receptor interacting complex protein, PRIC295, translational activator GCN1
Gene Symbols GCN1L1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2621 and 2671 of human general control of amino-acid synthesis 1-like 1 using the numbering given in entry NP_006827.1 (GeneID 10985).
Molecular Weight of Antigen 293 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10985
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.