Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCN2 Antibody, Novus Biologicals™
SDP

Catalog No. NB186424 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186424 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB186424 Supplier Novus Biologicals Supplier No. NBP312274
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chicken Polyclonal Antibody

GCN2 Polyclonal antibody specifically detects GCN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation

Specifications

Antigen GCN2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:200
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias eukaryotic translation initiation factor 2 alpha kinase 4, eukaryotic translation initiation factor 2-alpha kinase 4, GCN2GCN2 eIF2alpha kinase, GCN2-like protein, KIAA1338EC 2.7.11.1
Host Species Chicken
Immunogen Synthetic peptide taken within amino acid region 1600-1700 on human GCN2 protein
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 440275
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.