Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCN4 Antibody (C11L34), FITC, Novus Biologicals™
SDP

Catalog No. NB342791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342791 Supplier Novus Biologicals Supplier No. NBP281273F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GCN4 Monoclonal antibody specifically detects GCN4 in Yeast samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen GCN4
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone C11L34
Conjugate FITC
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Surface Plasmon Resonance
Formulation PBS
Gene Alias AAS101, AAS3, Amino acid biosynthesis regulatory protein, amino acid starvation-responsive transcription factor GCN4, ARG9, General Control Protein 4
Host Species Mouse
Immunogen Biotinylated GCN4-peptide (HLENEVARLKK).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856709
Target Species Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.