Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCN4 Antibody (C11L34), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB342782 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342782 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342782 Supplier Novus Biologicals Supplier No. NBP281274JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GCN4 Monoclonal antibody specifically detects GCN4 in Yeast samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen GCN4
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone C11L34
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Surface Plasmon Resonance
Formulation 50mM Sodium Borate
Gene Alias AAS101, AAS3, Amino acid biosynthesis regulatory protein, amino acid starvation-responsive transcription factor GCN4, ARG9, General Control Protein 4
Host Species Rabbit
Immunogen Biotinylated GCN4-peptide (HLENEVARLKK).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856709
Target Species Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.