Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCNF/NR6A1 Antibody (OTI6F3), PE, Novus Biologicals™
SDP

Catalog No. NB289350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289350 Supplier Novus Biologicals Supplier No. NBP270825PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GCNF/NR6A1 Monoclonal antibody specifically detects GCNF/NR6A1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen GCNF/NR6A1
Applications Western Blot
Classification Monoclonal
Clone OTI6F3
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias GCNF1, GCNFNR61, Germ cell nuclear factor, hGCNF, nuclear receptor subfamily 6 group A member 1, nuclear receptor subfamily 6, group A, member 1, retinoic acid receptor-related testis-associated receptor, Retinoid receptor-related testis-specific receptor, RTRhRTR
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-248 of human NR6A1(NP_001480) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Olfactory
Primary or Secondary Primary
Gene ID (Entrez) 2649
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.