Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCS1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP214967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214967 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP214967 Supplier Novus Biologicals Supplier No. NBP214967
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GCS1 Polyclonal specifically detects GCS1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockdown Validated.

Specifications

Antigen GCS1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence Reported in scientific literature (PMID:29465311) , Immunoprecipitation Validated from a verified customer review., Knockdown Validated Reported in scientific literature (PMID:29465311)
Formulation 0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q13724
Gene Alias GCS1, glucosidase I, mannosyl-oligosaccharide glucosidase, processing A-glucosidase I
Gene Symbols MOGS
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Glucosidase I. The exact sequence is proprietary.
Molecular Weight of Antigen 92 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7841
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.