Learn More
GCSAML Polyclonal Antibody

Description
GCSAML Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | GCSAML |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | GCSAML |
| Gene Accession No. | Q5JQS6 |
| Gene Alias | C1orf150; GCSAML; germinal center associated signaling and motility like; germinal center-associated signaling and motility-like protein; germinal center-associated, signaling and motility-like |
| Gene Symbols | GCSAML |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.