Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCSH Antibody (187), HRP, Novus Biologicals™
SDP

Catalog No. NB038584 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038584 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038584 Supplier Novus Biologicals Supplier No. NBP290290H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GCSH Monoclonal specifically detects GCSH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GCSH
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 187
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias GCE, glycine cleavage system H protein, mitochondrial, glycine cleavage system protein H (aminomethyl carrier), lipoic acid-containing protein, mitochondrial glycine cleavage system H-protein, NKH
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human GCSH (AAA36011.1; Ser49-Glu173).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2653
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.