Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB342242 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342242 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342242 Supplier Novus Biologicals Supplier No. NBP281826MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GDF-1 Polyclonal antibody specifically detects GDF-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen GDF-1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DORV, DTGA3, embryonic growth/differentiation factor 1, GDF-1, growth differentiation factor 1
Host Species Rabbit
Immunogen Rabbit polyclonal GDF-1 antibody was raised against a 16 amino acid peptide near the amino terminus of human GDF-1. The immunogen is located within amino acids 30 - 80 of GDF-1. Amino Acid Squence: CDEPQGAPRLRPVPPVM
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 2657
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.