Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-15 Antibody (7C12.B3.F2), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB120005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120005 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120005 Supplier Novus Biologicals Supplier No. NBP142725IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GDF-15 Monoclonal specifically detects GDF-15 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen GDF-15
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7C12.B3.F2
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta
Host Species Mouse
Immunogen This Protein A purified GDF-15 Antibody (7C12.B3.F2) was prepared by repeated immunizations with a synthetic peptide corresponding to a region near the amino terminal end of human GDF-15 protein. A residue of cysteine was added to facilitate coupling to KLH. (Uniprot: Q99988)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hepatic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9518
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.