Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-3 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB105292 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105292 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105292 Supplier Novus Biologicals Supplier No. NBP298719B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GDF-3 Polyclonal specifically detects GDF-3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GDF-3
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human GDF-3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9573
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.