Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-5/BMP-14 Antibody (RM0242-13J15), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP212333V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212333V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212333V Supplier Novus Biologicals Supplier No. NBP212333V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

GDF-5/BMP-14 Monoclonal specifically detects GDF-5/BMP-14 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen GDF-5/BMP-14
Applications Western Blot
Classification Monoclonal
Clone RM0242-13J15
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BMP14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2
Gene Symbols GDF5
Host Species Rat
Immunogen Mouse recombinant GDF-5/BMP-14
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 8200
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.