Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-8/Myostatin Antibody (GDF9/4261), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB391445 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391445 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391445 Supplier Novus Biologicals Supplier No. NBP308680FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GDF-8/Myostatin Monoclonal antibody specifically detects GDF-8/Myostatin in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen GDF-8/Myostatin
Applications Western Blot, ELISA
Classification Monoclonal
Clone GDF9/4261
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin
Host Species Mouse
Immunogen Recognizes the 113 amino acid carboxy-terminal fragment of GDF-8/Myostatin protein (Uniprot: O14793)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 2660
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.