Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-8/Myostatin Antibody (OTI7H5), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP270826IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270826IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270826IR Supplier Novus Biologicals Supplier No. NBP270826IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GDF-8/Myostatin Monoclonal specifically detects GDF-8/Myostatin in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen GDF-8/Myostatin
Applications Western Blot
Classification Monoclonal
Clone OTI7H5
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin
Gene Symbols MSTN
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 24-266 of human GDF-8/Myostatin (NP_005250) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 2660.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.