Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-9 Antibody (53/1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | GDF-9 |
---|---|
Clone | 53/1 |
Applications | ELISA, Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
GDF-9 | |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Mouse | |
PBS with 0.02% Sodium Azide | |
2661 | |
Tuberculin coupled peptide with sequence VPAKYSPLSVLTIEPDGSIAYKEYEDMIATKC that recognizes an epitope with the EPDG sequence near the C-terminal region of human GDF-9 | |
Primary |
53/1 | |
Monoclonal | |
Purified | |
Cytokine Research | |
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
GDF9 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title