Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-9 Antibody (72B), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP261933A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP261933A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP261933A70 Supplier Novus Biologicals Supplier No. NBP261933AF750

Rat Monoclonal Antibody

GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen GDF-9
Applications Western Blot, ELISA
Classification Monoclonal
Clone 72B
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9
Gene Symbols GDF9
Host Species Rat
Immunogen N-terminal of GDF-9, corresponding to sequence KKPLGPASFNLSEYFC
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 2661.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.