Learn More
Invitrogen™ GDF3 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ 500P235BT500UG

Description
AA Sequence of recombinant protein: AAIPVPKLSC KNLCHRHQLF INFRDLGWHK WIIAPKGFMA NYCHGECPFS LTISLNSSNY AFMQALMHAV DPEIPQAVCI PTKLSPISML YQDNNDNVIL RHYEDMVVDE CGCG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GDF-3. Anti-Human GDF-3-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hGDF-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human GDF-3 (500-P235) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hGDF-3. Western Blot: To detect hGDF-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hGDF-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P235BT-1MG will be provided as 2 x 500 μg
GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.
Specifications
GDF3 | |
Polyclonal | |
Biotin | |
GDF3 | |
C78318; ecat9; Gdf3; Gdf-3; growth differentiation factor 3; growth/differentiation factor 3; KFS3; MCOP7; MCOPCB6; RGD1564178; UNQ222/PRO248; VG-1-related protein 2; Vgr2; Vgr-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9573 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
0.1-1.0 mg/mL | |
PBS with no preservative | |
Q9NR23 | |
GDF3 | |
E.coli-derived Recombinant Human GDF-3. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.