Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDNF Antibody (MM0308-12F9), PerCP, Novus Biologicals™
SDP

Catalog No. NB361015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361015 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361015 Supplier Novus Biologicals Supplier No. NBP212335PCP

Mouse Monoclonal Antibody

GDNF Monoclonal antibody specifically detects GDNF in Human samples. It is validated for Western Blot, ELISA, Neutralization

Specifications

Antigen GDNF
Applications Western Blot, ELISA, Neutralization
Classification Monoclonal
Clone MM0308-12F9
Conjugate PerCP
Dilution Western Blot, ELISA, Block/Neutralize
Formulation PBS
Gene Alias Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3
Host Species Mouse
Immunogen Human recombinant GDNF
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2668
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.