Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDPD5 Antibody (506020) [Alexa Fluor™ 532], Novus Biologicals™
SDP

Catalog No. NB401445 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401445 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401445 Supplier Novus Biologicals Supplier No. FAB10524X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GDPD5 Monoclonal antibody specifically detects GDPD5 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen GDPD5
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 506020
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 3.1, GDE2PP1665Glycerophosphodiester phosphodiesterase 2, glycerophosphodiester phosphodiesterase domain containing 5, glycerophosphodiester phosphodiesterase domain-containing protein 5
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived transfected with GDPD5, Accession # Q8WTR4
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 81544
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.