Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDPD5 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343549 Supplier Novus Biologicals Supplier No. NBP241268MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GDPD5 Polyclonal antibody specifically detects GDPD5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen GDPD5
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 3.1, GDE2PP1665Glycerophosphodiester phosphodiesterase 2, glycerophosphodiester phosphodiesterase domain containing 5, glycerophosphodiester phosphodiesterase domain-containing protein 5
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the amino terminus of human GDPD5. The immunogen is located within the first 50 amino acids of GDPD5. Amino Acid Squence: RWKRYQRSHDDTTPWER
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 81544
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.