Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester)

Catalog No. 45000701 Shop All Cytiva Products
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Product Type:
Cy3 bis-NHS Ester
Cy5 Bis NHS Ester
Cy5 bis-NHS Ester
Cy5.5 Bis NHS Ester
Cy7 Bis NHS Ester

Suitable for general labeling of amine residues

Order Info

Large range of bulk packs available for greatest economy. The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Suitable for general labeling of Amine residues
Quantity 5 mg
Product Type Cy5 bis-NHS Ester
Kit Contents Foil packs, each containing 5, 10 or 50mg dried dye
Content And Storage 2°C to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.