Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva DeStreak Rehydration Solution

Catalog No. p-3667532 Shop All Cytiva Products
Click to view available options
Quantity:
1 mL
5 x 3 mL

Highly suitable for both analytical and preparative gels. DeStreak Rehydration Solution improves reproducibility and quality of 2-D gels by preventing streaking.

The appearance of streaks that distort 2-D electrophoresis maps is a common problem, occurring most frequently when running gels that contain regions greater than pH 7.0. Increased sample load, increased length of the IPG strip, or using a narrower pH gradient worsen the problem. Extra spots on 2-D gels, caused by nonspecific oxidation of proteins, is another difficulty encountered when running gels containing basic regions. Both streaking and nonspecific oxidation result in poorly resolved protein patterns and reduced reproducibility between electrophoresis runs. DeStreak Rehydration Solution effectively eliminates nonspecific oxidation by maintaining protein thiol groups in a single oxidation state, irrespective of sample load, pH range, or run length. Protein patterns are stabilized in IPG strips of any length and pH gradient ensuring the same stable and reproducible pattern in every analysis. The effectiveness of DeStreak Rehydration Solution is maintained at the high sample loads and long run times required for preparative gel electrophoresis.

  • Eliminates extra spots caused by nonspecific oxidation of proteins
  • Stabilizes proteins throughout entire 2-D runs, ensuring simplified, well-resolved protein patterns
  • Suitable for both analytical and preparative gels
  • Contains optimized concentrations of urea, thiourea, CHAPS, and DeStreak Reagent

Order Info

The solution is ready for use after addition of the appropriate IPG Buffer. DeStreak Reagent is also available separately in a 1mL pack size.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Improves reproducibility and quality of 2D Gels by preventing streaking
Quantity 1 mL
Product Type Destreak Reagent
Kit Contents Optimized concentrations of urea, thiourea, CHAPS and Destreak reagent
pH Range >7
Content And Storage 2°C to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.