Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva GST Bulk Kit
Shop All Cytiva Products
Click to view available options
:
Inc. 10mL Glutathione Sepharose 4B, 5 disposable columns
Description
- Gentle elution conditions preserve protein antigenicity and function
- Flexible GST-glutathione affinity system for rapid column or batch purification
- High specificity (compared to histidine-tagged purification)
- Provide reagents for induction expression and elution of fusion proteins
- Sufficient reagents to purify up to 50mg fusion protein
- Includes 10mL bulk pack of Glutathione Sepharose™ 4B, 5 disposible columns, GST buffer kit and 500 mg Isopropyl Β-D Thiogalactoside
Specifications
Specifications
| Description | GST bulk kit is designed for small-scale manual purification of GST-tagged proteins using either column chromatography or batch method. Facilitates optimization studies of expression, solubility conditions and purification parameters of GST-tagged protein |
| Kit Contents | 10 mL bulk pack of Glutathione Sepharose 4B medium, 5 x gravity-flow columns, IPTG, GST Buffer Kit |
| Content And Storage | 20°C to 25°C |
| Quantity | 1 Kit |
| Type | Chromatography Media |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction