Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Handbooks

Catalog No. 45002561 Shop All Cytiva Products
Click to view available options
Format:
Book

Catalog No. 45-002-561

Supplier: Cytiva 11001269

Increase knowledge of various subjects with these easy-to-follow handbooks

Includes additional reading, references and product ordering information.

TRUSTED_SUSTAINABILITY

Specifications

Format Book
Product Type Handbook of Hydrophobic Interaction and Reversed Phase Chromatography Principles and Methods
For Use With (Application) Describe both theoretical and practical aspects of the technique
Subject Biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.