Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GEF-H1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP221577 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221577 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221577 Supplier Novus Biologicals Supplier No. NBP221577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GEF-H1 Polyclonal specifically detects GEF-H1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.

Specifications

Antigen GEF-H1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000, Knockdown Validated
Formulation PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300
Gene Alias GEF, GEFH1, GEF-H1DKFZp547P1516, Guanine nucleotide exchange factor H1, KIAA0651DKFZp547L106, LFP40FLJ45440, Microtubule-regulated Rho-GEF, P40, Proliferating cell nucleolar antigen p40, rho guanine nucleotide exchange factor 2, Rho/Rac guanine nucleotide exchange factor (GEF) 2, Rho/Rac guanine nucleotide exchange factor 2
Gene Symbols ARHGEF2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human GEF-H1. The exact sequence is proprietary.
Molecular Weight of Antigen 112 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9181
Test Specificity Mouse reactivity determined by Western Blot.
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.