Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Geldanamycin
GREENER_CHOICE

Catalog No. 13681r
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective Hsp90 inhibitor

Selectively inhibits heat shock protein 90 (Hsp90). Binds to the ATP site of Hsp90 (Kd = 1.2 μM) and inhibits its chaperone activity. Consequently inhibits activities of oncogenic kinases (e.g. src, Raf), p53 and steroid receptors. Demonstrates antiproliferative effect on breast cancer stem-like cells.

Specifications

CAS 30562-34-6
Assay Percent Range >98%
Quantity 1 mg
Solubility Soluble to 10 mM in DMSO
Molecular Formula C29H40N2O9
Formula Weight 560.64g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.