Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelsolin/GSN Antibody (20), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP105161R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105161R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105161R Supplier Novus Biologicals Supplier No. NBP105161R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Gelsolin/GSN Monoclonal specifically detects Gelsolin/GSN in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Gelsolin/GSN
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 20
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type)
Gene Symbols GSN
Host Species Mouse
Immunogen Gelsolin purified from human plasma
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cytoskeleton Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2934
Test Specificity Gelsolin (human)
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.