Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelsolin/GSN Antibody, Novus Biologicals™
SDP

Catalog No. NBP227566 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227566 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227566 Supplier Novus Biologicals Supplier No. NBP227566
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

Gelsolin/GSN Polyclonal specifically detects Gelsolin/GSN in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Gelsolin/GSN
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000168.1
Gene Alias Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type)
Gene Symbols GSN
Host Species Goat
Immunogen Peptide with sequence PQGRVPEARPNS-C corresponding to N-Terminus according to NP_000168.1, NP_937895.1, NP_001121138.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cytoskeleton Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2934
Test Specificity This antibody is expected to recognize isoform a (NP_000168.1) only . Reported variants represent identical protein: NP_001121138.1, NP_001121139.1 Reported variants represent identical protein: NP_001121135.1, NP_937895.1, NP_001121134.1, NP_001121137.1, NP_001121136.1
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.