Learn More
Gelsolin Mouse, Unlabeled, Clone: 2, BD
Mouse Monoclonal Antibody
Specifications
Antigen | Gelsolin |
---|---|
Clone | 2 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB610412
|
BD Biosciences
610412 |
50 μg | N/A | N/A | N/A | ||||
Description
Gelsolin was identified as a result of its ability to sever actin filaments in a Ca2+-dependent manner. The gene for gelsolin encodes an 83 kDa protein that migrates as a 93 kDa polypeptide in SDS-gels. The N-terminal domain contains the calcium-independent actin-severin site, whereas the calcium-dependent site is located in the C-terminal portion of the protein. It exhibits significant homology with villin, another calcium-regulated actin filament severing protein. Gelsolin can be found intracellularly, as well as in a secreted form. However, both forms are encoded by the same gene.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
Gelsolin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
2 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rabbit, Rat | |
IgG2a | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.