Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GEMC1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB344025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344025 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344025 Supplier Novus Biologicals Supplier No. NBP241131MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GEMC1 Polyclonal antibody specifically detects GEMC1 in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen GEMC1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias GEMC1, Geminin Coiled-Coil Domain Containing, Geminin Coiled-Coil Domain-Containing Protein 1, GMNC
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the center of human GEMC1. The immunogen is located within amino acids 130 - 180 of GEMC1. Amino Acid Squence: YGKFRKGKRKSKEQRYSPA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 647309
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.