Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gemin 4 Antibody, Novus Biologicals™
SDP

Catalog No. NB11040591 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040591 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11040591 Supplier Novus Biologicals Supplier No. NB11040591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gemin 4 Polyclonal specifically detects Gemin 4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Gemin 4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. P57678
Gene Alias component of gems 4, DKFZp434B131, DKFZP434D174, gem (nuclear organelle) associated protein 4, gemin-4, HC56, HCAP1, HCC-associated protein 1, HHRF-1, p97DKFZp434D174
Gene Symbols GEMIN4
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1010 and the C-terminus (residue 1058) fo human Gem (nuclear organelle) Associated Protein 4 using the numbering given in entry NP_056536.1 (GeneID 50628).
Molecular Weight of Antigen 120 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50628
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, and Chimpanzee.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.