Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GEMIN2 Antibody (OTI6G4), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271375A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271375A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271375A48 Supplier Novus Biologicals Supplier No. NBP271375AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GEMIN2 Monoclonal specifically detects GEMIN2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen GEMIN2
Applications Western Blot
Classification Monoclonal
Clone OTI6G4
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Component of gems 2, Gemin-2, GEMIN2survival of motor neuron protein-interacting protein 1, SIP1-delta, SMN interacting protein 1-delta, SMN-interacting protein 1, survival of motor neuron protein interacting protein 1
Gene Symbols GEMIN2
Host Species Mouse
Immunogen Full length recombinant protein of human GEMIN2 (NP_003607) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8487.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.