Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Geminin Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7188616
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4142 10 μL
50-199-4141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4142 Supplier Active Motif Supplier No. 61136
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GMNN inhibits DNA replication by preventing the incorporation of MCM complex into prereplication complex (pre-RC). It is degraded during the mitotic phase of the cell cycle. Its destruction at the metaphase-anaphase transition permits replication in the succeeding cell cycle.

Specifications

Antigen Geminin
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene GMNN
Gene Accession No. O75496
Gene Alias AW546347; Gem; Geminin; Geminin GMNN; geminin, DNA replication inhibitor; Gmnn; MGORS6; OTTHUMP00000039393; RP3 369A17.3
Gene Symbols GMNN
Host Species Rabbit
Immunogen This Geminin antibody was raised against full-length recombinant human Geminin protein.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51053
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.