Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ GeneAmp™ 10X PCR Buffer

Catalog No. p-4919373
Encompass_Preferred
Click to view available options
Quantity:
1.5 mL
6 x 1.5 mL

Applied Biosystems GeneAmp 10X PCR Buffer I is optimized for use with AmpliTaq DNA Polymerase, resulting in robust PCR amplification.

Applied Biosystems™ GeneAmp™ 10X PCR Buffer I is optimized for use with AmpliTaq™ DNA Polymerase, resulting in robust PCR amplification.

  • Optimized for use with AmpliTaq™ DNA Polymerase, offering maximum PCR performance
  • Available in several convenient sizes

GeneAmp™ 10X PCR Buffer I contains 15 mM MgCl2. When diluted 1:10, it provides the optimal pH and ionic strength for PCR amplification with AmpliTaq™ DNA Polymerase.

Note: See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material PCR Buffer
Concentration 10X
For Use With (Application) Standard PCR
Quantity 1.5 mL
Includes • GeneAmp 10X PCR Buffer I (with 15 mM MgCl2), 1.5 mL

Store at -15°C to -25°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.