Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ GeneAmp™ 10X PCR Gold Buffer & MgCl2

Applied Biosystems GeneAmp 10X PCR Gold Buffer is formulated to provide flexible, efficient activation of AmpliTaq Gold DNA Polymerase, resulting in highly specific, robust PCR amplification.

$143.00 - $315.00

Specifications

Concentration 10X
Format Kit
Polymerase AmpliTaq Gold DNA Polymerase
Product Type 10X PCR Gold Buffer
For Use With (Application) Standard PCR
View More Specs

Products 2
Catalog Number Mfr. No. Volume Price Quantity & Availability  
Catalog Number Mfr. No. Volume Price Quantity & Availability  
43-798-74
Encompass_Preferred
View Documents
Applied Biosystems™
4379874
2 x 75 mL
Each for $315.00
Only null left
Add to Cart
 
43-068-98
Encompass_Preferred
View Documents
Applied Biosystems™
4306898
6 x 1.5 mL
Each for $143.00
Only null left
Add to Cart
 
Description

Description

Applied Biosystems GeneAmp 10X PCR Gold Buffer and MgCl2 is formulated to provide flexible, efficient activation of AmpliTaq Gold DNA Polymerase, resulting in highly specific, robust PCR amplification. The ionic strength and pH of GeneAmp 10X PCR Gold Buffer has been optimized to provide a wider activation temperature and time range when used in with AmpliTaq Gold DNA Polymerase.

When GeneAmp 10X PCR Gold Buffer and the MgCl2 solution are used together, there is greater versatility in optimizing magnesium ion concentrations for optimal PCR amplification with any specific set of primers and DNA template.

Features

  • Magnesium ion–free
  • Gelatin-free
  • High versatility in optimizing magnesium ion concentration
  • MgCl2 solution provided separately
Specifications

Specifications

10X
AmpliTaq Gold DNA Polymerase
Standard PCR
Kit
10X PCR Gold Buffer
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.