Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Precision gRNA Synthesis Kit

Catalog No. a29377
Encompass_Preferred
Click to view available options
Quantity:
25 reactions
For research use only. Not for use in diagnostic procedures.
For research use only. Not for use in diagnostic procedures.

Includes

BOX 1

  • 5X TranscriptAidTM Reaction Buffer
  • DNase I, RNase-free (1 U/μL)
  • Nuclease-free water
  • NTP mix (100 mM each of ATP, GTP, CTP,
  • UTP in Tris buffer)
  • Control gRNA forward and reverse primers (10 μM mix) 2

BOX 2

  • Binding Buffer
  • Wash Buffer 1 (concentrated)3
  • Wash Buffer 2 (concentrated)4
  • Nuclease-free water
  • Gene JETTM RNA Purification Micro Column & Collection Tubes
  • Elution Tubes, 1.5 mL

Simple, ready-to-use, all-in-one expression vector system for CRISPR-Cas9. Transfection-ready guide RNA (gRNA) in as little as four hours.

Simple, ready-to-use, all-in-one expression vector system for CRISPR-Cas9. GeneArt Precision gRNA Synthesis Kit is a complete system for rapid synthesis of guide RNA (gRNA) ready to complex with GeneArt Platinum Cas9 Nuclease or GeneArt CRISPR Nuclease mRNA. Starting with two short single-stranded oligos that code for the target sequence, the gRNA template is assembled with a T7 promoter in a short one-pot PCR reaction. The assembled product is then used as template in an in vitro transcription (IVT) reaction followed by a rapid purification step, yielding transfection-ready gRNA in as little as four hours. Resulting gRNA can also be co-transfected with our ready-to-transfect GeneArt Platinum Cas9 Nuclease or GeneArt CRISPR Nuclease mRNA. Both protein and mRNA Cas9 formats require no plasmid manipulation and so are amenable to high throughput and multiplex genome-wide cell engineering approaches.

Features of the GeneArt Precision gRNA Synthesis Kit include:

  • Fast assembly and synthesis of any gRNA target in as little as four hours including template assembly
  • High yield (>10 ug) and concentration (>200 ng/uL) of gRNA

For research use only. Not for use in diagnostic procedures.

Certifications

CoA

Recommended Storage

gRNA Prep Kit, store at -5 to -30°C and gRNA Cleanup Kit, store at room temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 200 ng/μL
Content And Storage • gRNA Prep Kit, store at -5°C to -30°C
• - gRNA Cleanup Kit, store at room temperature (For better long-term performance, store purification columns at 2°C to 8°C)
Form Liquid
Format Kit
Reaction Speed Fast
Technique CRISPR-Cas9
Product Type gRNA Synthesis Kit
Promoter T7
Quantity 25 reactions
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Sufficient For 25 Reactions
Yield >10 μg at >200 ng/μL of gRNA
Final Product Type RNA
No. of Reactions 25 Reactions
Starting Material Oligos (DNA)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.